missing translation for 'onlineSavingsMsg'
Learn More
4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals Product Code.: 11916301

4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals

Product Code. 11916301
2.5 kg, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 11916301

Brand: Thermo Scientific Acros 148270025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-58-3
MDL Number MFCD00006418
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
Synonym 4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamineShow MoreShow Less
PubChem CID 14284
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)C1=CC=NC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 110.0°C to 113.0°C
Boiling Point 162.0°C (50.0 mmHg)
Flash Point 124°C
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Molecular Formula C7H10N2
Linear Formula (C5H4N)N(CH3)2
Quantity 2.5 kg
Beilstein 22, II, 341
Fieser 03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142
Merck Index 15, 3433
Solubility Information Solubility in water: 76g/L (25°C). Other solubilities: soluble in ether, slightly soluble in alcohol, benzene and, chloroform
Molecular Weight (g/mol) 122.17
Formula Weight 122.17
Vapor Pressure Negligible
Percent Purity 99%
Chemical Name or Material 4-Dimethylaminopyridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454276 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number148275000,148270250,148270050,148271000,148270025
Lot NumberA0465095 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number148275000,148270250,148270050,148271000,148270025
Lot NumberA0459233 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number148275000,148270250,148270050,148271000,148270025
Lot NumberA0440482 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number148275000,148270250,148270050,148271000,148270025
Lot NumberA0454276 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number148275000,148270250,148270050,148271000,148270025
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Dimethylaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.