missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylsulfamoyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15409707
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486422-59-7
Molecular Formula C8H12BNO4S
Molecular Weight (g/mol) 229.06
MDL Number MFCD06659849
InChI Key VVUGXBSXXJTVDS-UHFFFAOYSA-N
Synonym 4-n,n-dimethylsulfamoyl phenylboronic acid, n,n-dimethyl 4-boronobenzenesulfonamide, 4-n,n-dimethylsulphonamido benzeneboronic acid, 4-dimethylsulfamoyl phenylboronic acid, 4-dimethylsulfamoyl phenyl boronic acid, 4-n,n-dimethylsulfamoyl phenyl boronic acid, 4-borono-n,n-dimethylbenzenesulphonamide, 4-n,n-dimethylsulfonamidophenyl boronic acid, n,n-dimethyl-4-boronobenzenesulfonamide, boronic acid, 4-dimethylamino sulfonyl phenyl
PubChem CID 44119328
IUPAC Name [4-(dimethylsulfamoyl)phenyl]boronic acid
SMILES CN(C)S(=O)(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 140°C to 144°C
Quantity 1 g
Formula Weight 229.06
Percent Purity 95%
Chemical Name or Material 4-(Dimethylsulfamoyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'