missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylamino)benzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 15442289
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. Afficher la politique du retour.

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 4181-05-9
Molecular Formula C19H15NO
Molecular Weight (g/mol) 273.34
MDL Number MFCD00145131
InChI Key UESSERYYFWCTBU-UHFFFAOYSA-N
Synonym 4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
PubChem CID 77846
IUPAC Name 4-(N-phenylanilino)benzaldehyde
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O

Spécification

Melting Point 129°C to 133°C (literature)
Color Yellow
Odor Odorless
Quantity 5 g
Beilstein 2732795
Sensitivity Air sensitive
Solubility Information Soluble in methanol.
Formula Weight 273.33
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Diphenylamino)benzaldehyde
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'