missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[(Dipropylamino)sulfonyl]benzoic acid, 97%, Maybridge™
£21.00 - £67.00
Chemical Identifiers
CAS | 57-66-9 |
---|---|
Molecular Formula | C13H19NO4S |
Molecular Weight (g/mol) | 285.36 |
MDL Number | MFCD00038402 |
InChI Key | DBABZHXKTCFAPX-UHFFFAOYSA-N |
Synonym | probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin |
PubChem CID | 4911 |
ChEBI | CHEBI:8426 |
IUPAC Name | 4-(dipropylsulfamoyl)benzoic acid |
SMILES | CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O |
Description
Chemical Identifiers
57-66-9 | |
285.36 | |
DBABZHXKTCFAPX-UHFFFAOYSA-N | |
4911 | |
4-(dipropylsulfamoyl)benzoic acid |
C13H19NO4S | |
MFCD00038402 | |
probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin | |
CHEBI:8426 | |
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O |
Specifications
57-66-9 | |
97% | |
[CH3(CH2)2]2NSO2C6H4CO2H | |
probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin | |
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O | |
285.36 | |
CHEBI:8426 | |
97% |
97.0 | |
C13H19NO4S | |
MFCD00038402 | |
DBABZHXKTCFAPX-UHFFFAOYSA-N | |
4-(dipropylsulfamoyl)benzoic acid | |
4911 | |
285.36 | |
4-[(Dipropylamino)sulfonyl]benzoic acid |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning