missing translation for 'onlineSavingsMsg'
Learn More

4-Dodecyldiethylenetriamine, 80%, tech., Thermo Scientific Chemicals

Product Code. 10659964
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4182-44-9
CAS Min % 75.0
CAS Max % 100.0
Melting Point 22°C
Color Colorless to Yellow
Boiling Point 140°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Molecular Formula C16H37N3
Linear Formula CH3(CH2)11N(CH2CH2NH2)2
Refractive Index 1.4640 to 1.468
MDL Number MFCD00041897
Quantity 25 g
Synonym 4-dodecyldiethylenetriamine, unii-vg94uin7ys, 1,2-ethanediamine, n-2-aminoethyl-n-dodecyl, n-2-aminoethyl-n-dodecyl-1,2-ethanediamine, vg94uin7ys, n1-2-aminoethyl-n1-dodecyl-1,2-ethanediamine, 1,2-ethanediamine, n1-2-aminoethyl-n1-dodecyl, n-2-aminoethyl-n-dodecylethylenediamine, bis 2-aminoethyl dodecyl amine, n'-2-aminoethyl-n'-dodecylethane-1,2-diamine
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,chloroform,benzene,toluene,,diethyl ether
InChI Key BRNPMBFYMPUNSK-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCN(CCN)CCN
IUPAC Name N'-(2-aminoethyl)-N'-dodecylethane-1,2-diamine
Molecular Weight (g/mol) 271.48
PubChem CID 77847
Formula Weight 271.48
Percent Purity 80%
Physical Form Liquid
Chemical Name or Material 4-Dodecyldiethylenetriamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Dodecyldiethylenetriamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'