missing translation for 'onlineSavingsMsg'
Learn More

4-Epianhydrochlortetracycline hydrochloride, can be used as secondary standard, Thermo Scientific Chemicals

Código de producto. 10308960 Tienda Thermo Scientific Chemicals Productos
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
missing translation for 'unitSize'
100mg
500mg
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10308960

missing translation for 'mfr': Thermo Scientific Acros 268225000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158018-53-2
Molecular Formula C22H21ClN2O7·HCl
Molecular Weight (g/mol) 497.33
MDL Number MFCD00143313
InChI Key ISGAAFMBTIWTEU-PXAZKYFKSA-N
Synonym 4-epianhydrochlortetracycline hydrochloride, 4-epianhydrochlortetracycline hcl, epianhydrochlortetracycline hydrochloride, 2z,4r,4as,12as-2-amino hydroxy methylidene-7-chloro-4-dimethylamino-10,11,12a-trihydroxy-6-methyl-4a,5-dihydro-4h-tetracene-1,3,12-trione hydrochloride, 4r,4as,12ar-7-chloro-4-dimethylamino-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide-hydrogen chloride 1/1, 4r,4as,12as-7-chloro-4-dimethylamino-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-4a,5-dihydro-4h-tetracene-2-carboxamide hydrochloride, 4r-4, a,4a, a,12a, a-7-chloro-4-dimethylamino-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride
PubChem CID 54710413
IUPAC Name (4R,4aS,12aR)-7-chloro-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C14)Cl)O)O)O)O)C(=O)N)N(C)C.Cl

Specifications

Melting Point 222.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: 10g/L. Other solubilities: 29g/L methanol
Formula Weight 497.33
Percent Purity ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Physical Form Powder
Chemical Name or Material 4-Epianhydrochlortetracycline hydrochloride, Can be used as secondary standard
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Epianhydrochlortetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.