missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 4-Epitetracycline hydrochloride, can be used as a secondary standard Product Code.: 10318370

Thermo Scientific Chemicals 4-Epitetracycline hydrochloride, can be used as a secondary standard

Product Code. 10318370
500 mg, Glass bottle
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10318370

Brand: Thermo Scientific Chemicals 233125000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Epitetracycline hydrochloride, CAS # 23313-80-6, is a degradation product of tetracycline that shows antibiotic activity and can induce the protein biosynthesis inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 4-Epitetracycline hydrochloride is a metabolite derived from the degradation of tetracycline compounds and has been shown to display antibiotic activity
  • This compound blocks protein synthesis by binding to the 30S ribosomal subunit and interfering with the binding of aminoacyl-tRNA to the mRNA-ribosome complex

Applications

  • This semi-synthesized compound derived from actinomycetes can decrease the activity of the Tet repressor effector in animal models
  • In vivo experiments revealed that 4-epitetracycline hydrochloride can induce a toxic response
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23313-80-6
Molecular Formula C22H24N2O8·HCl
Molecular Weight (g/mol) 480.89
MDL Number MFCD00865028
InChI Key YCIHPQHVWDULOY-DXDJYCPMSA-N
Synonym 4-epitetracycline hydrochloride, epitetracycline hydrochloride, 4-epi-tetracycline hydrochloride, quatrimycin hydrochloride, epitetracycline hcl, unii-i7c3edl2yg, i7c3edl2yg, epitetracycline hydrochloride usp, 4r,4as,5as,6s,12as-4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochlorideShow MoreShow Less
PubChem CID 54686189
IUPAC Name (4R,4aS,5aS,6S,12aR)-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochlorideShow MoreShow Less
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Melting Point 217.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Quantity 500 mg
Specific Rotation − 322.00
Solubility Information Solubility in water: 100g/L. Other solubilities: 100g/L methanol
Formula Weight 480.89
Percent Purity ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Epitetracycline hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462250 Certificate TypeCertificate of Analysis Date13/05/2024 Catalog Number233125000,233121000
Lot NumberA0456458 Certificate TypeCertificate of Analysis Date26/09/2023 Catalog Number233125000,233121000
Lot NumberA0395930 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number233125000,233121000
Lot NumberA0400799 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number233125000,233121000
Lot NumberA0397675 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number233125000,233121000
compliance-icons compliance-icons
Product Identifier
  • 4-Epitetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 4-Epitetracycline hydrochloride, can be used as a secondary standard >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.