missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-1,1,1-trifluoro-3-buten-2-one, 97%

Product Code. 11964881 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11964881

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17129-06-5 | C6H7F3O2 | 168.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 17129-06-5
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1800g/mL
Boiling Point 51.0°C to 53.0°C (12.0 mmHg)
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C6H7F3O2
MDL Number MFCD00192131
Quantity 5 g
Specific Gravity 1.18
Synonym 4-ethoxy-1,1,1-trifluoro-3-buten-2-one, 4-ethoxy-1,1,1-trifluorobut-3-en-2-one, e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, 3e-4-ethoxy-1,1,1-trifluorobut-3-en-2-one, etfbo, 2-ethoxyvinyl trifluoromethyl ketone, e-4-ethoxy-1,1,1-trifluoro-3-buten-2-one, 3-buten-2-one, 4-ethoxy-1,1,1-trifluoro, 4-ethoxy, 4-ethoxy-1,1,1-trifluoro-but-3-en-2-one
InChI Key YKYIFUROKBDHCY-UHFFFAOYSA-N
SMILES CCOC=CC(=O)C(F)(F)F
IUPAC Name (E)-4-ethoxy-1,1,1-trifluorobut-3-en-2-one
Stabilizer (0.5% BHT stabilizer)
Molecular Weight (g/mol) 168.12
PubChem CID 5709222
Formula Weight 168.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Ethoxy-1, 1, 1-trifluoro-3-buten-2-one
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Ethoxy-1,1,1-trifluoro-3-buten-2-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.