missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,3-difluorophenol, 97%, Thermo Scientific™

Product Code. 15474697
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126163-56-2
Molecular Formula C8H8F2O2
Molecular Weight (g/mol) 174.147
MDL Number MFCD07368025
InChI Key PKIYFPSPIFCDDB-UHFFFAOYSA-N
Synonym 1-hydroxy-4-ethoxy-2,3-difluorobenzene, phenol, 4-ethoxy-2,3-difluoro, 2,3-difluoro-4-ethoxyphenol, 4-ethoxy-2,3-difluorophenol, jrd
PubChem CID 14495763
IUPAC Name 4-ethoxy-2,3-difluorophenol
SMILES CCOC1=C(C(=C(C=C1)O)F)F

Specifications

Melting Point 65°C to 77°C
Quantity 1 g
Formula Weight 174.14
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,3-difluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'