missing translation for 'onlineSavingsMsg'
Learn More
4-Ethoxy-2-methylbenzeneboronic acid, 98%, Thermo Scientific™ Product Code.: 15429277

4-Ethoxy-2-methylbenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15429277
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15429277

Brand: Thermo Scientific Alfa Aesar H52538.03

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313545-31-2
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD04115659
InChI Key MBEQXBKSJCOKJF-UHFFFAOYSA-N
Synonym 4-ethoxy-2-methylbenzeneboronic acid, 4-ethoxy-2-methylphenylboronicacid, 4-ethoxy-2-methylphenyl boronic acid, 5-ethoxytoluene-2-boronic acid, 4-ethoxy-2-methyl-phenyl boronic acid, boronic acid,b-4-ethoxy-2-methylphenyl, acmc-1aexl, 2-methyl-4-ethoxybenzene boronic acid, contains varying amounts of anhydride
PubChem CID 16217476
IUPAC Name (4-ethoxy-2-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OCC)C)(O)O

Specifications

Melting Point 163°C to 168°C
Quantity 1 g
Formula Weight 180.01
Percent Purity 98%
Chemical Name or Material 4-Ethoxy-2-methylbenzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB-BG280502 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Ethoxy-2-methylbenzeneboronic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.