missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2-methylbenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15439277
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313545-31-2
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD04115659
InChI Key MBEQXBKSJCOKJF-UHFFFAOYSA-N
Synonym 4-ethoxy-2-methylbenzeneboronic acid, 4-ethoxy-2-methylphenylboronicacid, 4-ethoxy-2-methylphenyl boronic acid, 5-ethoxytoluene-2-boronic acid, 4-ethoxy-2-methyl-phenyl boronic acid, boronic acid,b-4-ethoxy-2-methylphenyl, acmc-1aexl, 2-methyl-4-ethoxybenzene boronic acid, contains varying amounts of anhydride
PubChem CID 16217476
IUPAC Name (4-ethoxy-2-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OCC)C)(O)O

Specifications

Melting Point 163°C to 168°C
Quantity 5 g
Formula Weight 180.01
Percent Purity 98%
Chemical Name or Material 4-Ethoxy-2-methylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'