missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 15407287
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1206593-33-0
Molecular Formula C10H11F3O2
Molecular Weight (g/mol) 220.191
MDL Number MFCD13196581
InChI Key SRVUDCQCZVEMAK-UHFFFAOYSA-N
Synonym 4-ethoxy-3-trifluoromethyl benzyl alcohol, 4-ethoxy-3-trifluoromethyl phenyl methanol, 4-ethoxy-3-trifluoromethyl phenyl methan-1-ol
PubChem CID 57361412
IUPAC Name [4-ethoxy-3-(trifluoromethyl)phenyl]methanol
SMILES CCOC1=C(C=C(C=C1)CO)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 233.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Ethoxy-3-(trifluoromethyl)benzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'