missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™

Product Code. 15488785
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1206594-24-2
Molecular Formula C12H11F3O3
Molecular Weight (g/mol) 260.212
MDL Number MFCD13196588
InChI Key WNOQTODUBHURAA-GQCTYLIASA-N
Synonym 2e-3-4-ethoxy-3-trifluoromethyl phenyl prop-2-enoic acid, 3-4-ethoxy-3-trifluoromethyl phenyl acrylic acid
PubChem CID 60138059
IUPAC Name (E)-3-[4-ethoxy-3-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES CCOC1=C(C=C(C=C1)C=CC(=O)O)C(F)(F)F

Specifications

Quantity 250 mg
Formula Weight 260.21
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-3-(trifluoromethyl)cinnamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Ethoxy-3-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.