missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl trans-4-(4-n-pentylcyclohexyl)benzoate, 99%, Thermo Scientific™

Product Code. 15472726
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95973-51-6
Molecular Formula C26H34O3
Molecular Weight (g/mol) 394.555
MDL Number MFCD09952999
InChI Key BOJBRLBJBXWHNZ-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl 4-trans-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl 2-trans-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl 4-1s,4r-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl 4-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl-4'-trans-pentylcyclohexyl benzoate, 4-ethoxyphenyl trans-4-4-n-pentylcyclohexyl benzoate
PubChem CID 23334669
IUPAC Name (4-ethoxyphenyl) 4-(4-pentylcyclohexyl)benzoate
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OC3=CC=C(C=C3)OCC

Specifications

Quantity 1 g
Formula Weight 394.55
Percent Purity 99%
Chemical Name or Material 4-Ethoxyphenyl trans-4-(4-n-pentylcyclohexyl)benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.