missing translation for 'onlineSavingsMsg'
Learn More
4-Ethylbenzoyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 10093740

4-Ethylbenzoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 10093740
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10093740

Brand: Thermo Scientific Acros 305730050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16331-45-6
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000697
InChI Key AVTLLLZVYYPGFX-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl
PubChem CID 85368
IUPAC Name 4-ethylbenzoyl chloride
SMILES CCC1=CC=C(C=C1)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1400g/mL
Boiling Point 235°C to 236°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5C6H4COCl
Refractive Index 1.5465 to 1.5485
Quantity 5 g
Specific Gravity 1.14
Formula Weight 168.62
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Ethylbenzoyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.