missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzoyl chloride, 98+%, Thermo Scientific Chemicals

Product Code. 11324739
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16331-45-6
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000697
InChI Key AVTLLLZVYYPGFX-UHFFFAOYSA-N
PubChem CID 85368
IUPAC Name 4-ethylbenzoyl chloride
SMILES CCC1=CC=C(C=C1)C(=O)Cl

Specifications

Boiling Point 235°C to 236°C
Flash Point >110°C (230°F)
Odor Acidic
Quantity 5 g
UN Number UN3265
Beilstein 2206041
Synonym benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl
Formula Weight 168.62
Percent Purity ≥98%
Chemical Name or Material 4-Ethylbenzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'