missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylbenzyl chloride, 97%, stab. with calcium carbonate, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.04 |
Boiling Point | 111°C (26 mmHg) |
Quantity | 5 g |
UN Number | UN3265 |
Solubility Information | Insoluble in water. |
Formula Weight | 154.64 |
Percent Purity | 97% |
Chemical Name or Material | 4-Ethylbenzyl chloride, stabilized with calcium carbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'