missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylmorpholine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
Description
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.
Solubility
Miscible with water.
Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.
Solubility
Miscible with water.
Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -63°C |
Density | 0.908 |
Boiling Point | 138°C to 139°C |
Flash Point | 27°C |
Refractive Index | 1.4413 |
Quantity | 250 mL |
UN Number | UN2734 |
Beilstein | 102969 |
Sensitivity | Air Sensitive |
Synonym | n-ethylmorpholine, morpholine, 4-ethyl |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'