missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylphenol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1000 g
Description
Applications
4-Ethylphenol is suitable for use in the determination of 4-ethylguaiacol and 4-ethylphenol in the aroma of red wines using headspace-solid-phase microextraction method. It is a chemical reagent used in the synthesis of 5-HT1A recep
4-Ethylphenol is suitable for use in the determination of 4-ethylguaiacol and 4-ethylphenol in the aroma of red wines using headspace-solid-phase microextraction method. It is a chemical reagent used in the synthesis of 5-HT1A recep
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 123-07-9 |
Melting Point | 40°C to 45°C |
Density | 1.011 |
Boiling Point | 218°C to 219°C |
Flash Point | 100°C |
Molecular Formula | C8H10O |
Refractive Index | 1.533 |
MDL Number | MFCD00002393 |
Quantity | 1000 g |
UN Number | UN2430 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'