missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenyl isocyanate, 98%

Product Code. 11343336
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11343336

Brand: Thermo Scientific Alfa Aesar L12718.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23138-50-3 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23138-50-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00013881
InChI Key FWPYUSLQCQDLJR-UHFFFAOYSA-N
PubChem CID 90007
IUPAC Name 1-ethyl-4-isocyanatobenzene
SMILES CCC1=CC=C(C=C1)N=C=O

Specifications

Density 1.024
Boiling Point 67°C to 68°C (1 mmHg)
Flash Point 78°C (172°F)
Odor Pungent
Refractive Index 1.527
Quantity 5 g
UN Number UN2206
Beilstein 971100
Sensitivity Moisture sensitive
Synonym 4-ethylphenyl isocyanate, p-ethylphenyl isocyanate, 4-ethylphenylisocyanate, benzene, 1-ethyl-4-isocyanato, benzene,1-ethyl-4-isocyanato, 4-ethylbenzenisocyanate, acmc-1cpzx, 4-ethyl phenylisocyanate, 4-ethyl-phenyl-isocyanate, 1-ethyl-4-isocyanato-benzene
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 4-Ethylphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.