missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific™

Product Code. 15490806
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Ethylphenylmagnesium bromide, 0.5M in 2-MeTHF is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidi
TRUSTED_SUSTAINABILITY

Specifications

CAS 22873-28-5
Molecular Formula C8H9BrMg
MDL Number MFCD01311467
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 4-ethylphenylmagnesium bromide, 4-ethylphenyl magnesium bromide, vlouakpiqasfds-uhfffaoysa-m, 4-ethylphenylmagnesium bromide, 0.5m in 2-methf, 4-ethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 4-ethylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Reacts with water.
InChI Key QSGSHZBZGXYVOQ-UHFFFAOYSA-M
SMILES [Mg++].[Br-].CCC1=CC=[C-]C=C1
IUPAC Name magnesium;ethylbenzene;bromide
Molecular Weight (g/mol) 209.37
PubChem CID 11241259
Formula Weight 209.36
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 4-Ethylphenylmagnesium bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'