missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-1,3-dioxolan-2-one, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 18°C to 23°C |
Color | Colorless |
Boiling Point | 212°C |
Flash Point | >102°C (216°F) |
Quantity | 100 g |
Solubility Information | Slightly miscible with water. |
Formula Weight | 106.05 |
Percent Purity | 98% |
Physical Form | Liquid |
Chemical Name or Material | 4-Fluoro-1,3-dioxolan-2-one |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'