missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-2-methoxybenzylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
As pharmaceutical intermediate. As a metabolite of Capsaicin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
As pharmaceutical intermediate. As a metabolite of Capsaicin.
Solubility
Insoluble in water.
Notes
Air Sensitive and Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
As pharmaceutical intermediate. As a metabolite of Capsaicin.
Solubility
Insoluble in water.
Notes
Air Sensitive and Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications
Specifications
Refractive Index | 1.524 |
Quantity | 1 g |
UN Number | UN2735 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 155.17 |
Percent Purity | 97% |
Chemical Name or Material | 4-Fluoro-2-methoxybenzylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'