missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 11474693
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-71-1
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007832
InChI Key KMHLGVTVACLEJE-UHFFFAOYSA-N
Synonym 2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene
PubChem CID 67982
IUPAC Name 4-fluoro-2-methylaniline
SMILES CC1=CC(F)=CC=C1N

Specifications

Boiling Point 90°C to 92°C (16 mmHg)
Flash Point 87°C (188°F)
Odor Characteristic
Quantity 5 g
UN Number UN2810
Beilstein 1931798
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'