Learn More
4-Fluoro-2-methylaniline, 97%, Thermo Scientific Chemicals
£21.40 - £65.50
Chemical Identifiers
CAS | 452-71-1 |
---|---|
Molecular Formula | C7H8FN |
Molecular Weight (g/mol) | 125.15 |
MDL Number | MFCD00007832 |
InChI Key | KMHLGVTVACLEJE-UHFFFAOYSA-N |
Synonym | 2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene |
PubChem CID | 67982 |
IUPAC Name | 4-fluoro-2-methylaniline |
SMILES | CC1=CC(F)=CC=C1N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474693
|
Thermo Scientific Alfa Aesar
A11609.06 |
5 g |
£21.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484693
|
Thermo Scientific Alfa Aesar
A11609.14 |
25 g |
£65.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
452-71-1 | |
125.15 | |
KMHLGVTVACLEJE-UHFFFAOYSA-N | |
67982 | |
CC1=CC(F)=CC=C1N |
C7H8FN | |
MFCD00007832 | |
2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene | |
4-fluoro-2-methylaniline |
Specifications
452-71-1 | |
87°C (188°F) | |
C7H8FN | |
UN2810 | |
2-amino-5-fluorotoluene, benzenamine, 4-fluoro-2-methyl, 4-fluoro-2-methylbenzenamine, 6-amino-3-fluorotoluene, o-toluidine, 4-fluoro, 2-methyl-4-fluoroaniline, 4-fluorotoluidine, 4-fluoro-o-toluidine, 3-fluoro-6-aminotoluene | |
CC1=CC(F)=CC=C1N | |
125.15 | |
125.15 | |
4-Fluoro-2-methylaniline |
90°C to 92°C (16 mmHg) | |
Characteristic | |
MFCD00007832 | |
1931798 | |
KMHLGVTVACLEJE-UHFFFAOYSA-N | |
4-fluoro-2-methylaniline | |
67982 | |
97% |
Safety and Handling
GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P210-P260-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : CY0500000
missing translation for 'tsca' : TSCA
RUO – Research Use Only