missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11338886
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7079-48-3
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD03094390
InChI Key XLPGWKNCWMFHOD-UHFFFAOYSA-N
Synonym 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl
PubChem CID 2778602
IUPAC Name 4-fluoro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)F)S(=O)(=O)Cl

Specifications

Density 1.433
Boiling Point 245°C to 246°C
Flash Point >110°C (230°F)
Refractive Index 1.537
Quantity 1 g
UN Number UN3265
Beilstein 6720115
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'