missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenol, 98%, Thermo Scientific Chemicals

Product Code. 11458676
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-72-2
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00075088
InChI Key GKQDDKKGDIVDAG-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
PubChem CID 136295
IUPAC Name 4-fluoro-2-methylphenol
SMILES CC1=C(C=CC(=C1)F)O

Specifications

Melting Point 32°C to 36°C
Boiling Point 87°C (14 mmHg)
Flash Point 85°C (185°F)
Odor Phenol-like
Refractive Index 1.5165
Quantity 100 g
UN Number UN3261
Beilstein 2041490
Solubility Information Insoluble in water.
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-methylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'