missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10003393
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 139911-29-8
MDL Number MFCD02093072
InChI Key IQMLIVUHMSIOQP-UHFFFAOYSA-N
Synonym 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp
PubChem CID 2734665
IUPAC Name (4-fluoro-2-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)F)C)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 194°C to 196°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H8BFO2
Quantity 25 g
Molecular Weight (g/mol) 153.95
Formula Weight 153.95
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Fluoro-2-methylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-methylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.