missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11364729
Click to view available options
Quantity:
50 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-78-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007830
InChI Key PUGDHSSOXPHLPT-UHFFFAOYSA-N
Synonym 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
PubChem CID 67769
IUPAC Name 4-fluoro-2-nitroaniline
SMILES NC1=CC=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 90°C to 94°C
Flash Point 89°C (192°F)
Quantity 50 g
Beilstein 2210197
Solubility Information Insoluble in water.
Formula Weight 156.12
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-nitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'