missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15436727
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1323966-38-6
Molecular Formula C8H3ClF4O2
Molecular Weight (g/mol) 242.55
MDL Number MFCD19687189
InChI Key RAPKOVZLHNOHEJ-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethoxy benzoyl chloride
PubChem CID 66523624
IUPAC Name 4-fluoro-2-(trifluoromethoxy)benzoyl chloride
SMILES FC1=CC(OC(F)(F)F)=C(C=C1)C(Cl)=O

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 242.55
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethoxy)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'