missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 11498937
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208173-21-1
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD00061251
InChI Key WBCCAINPZLAKRN-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethanone, 4-fluoro-2-trifluoromethyl acetophenone, 1-4-fluoro-2-trifluoromethyl phenyl ethan-1-one, ethanone, 1-4-fluoro-2-trifluoromethyl phenyl, 1-acetyl-4-fluoro-2-trifluoromethyl benzene, pubchem4243, acmc-1cmiy, 2-acetyl-5-fluorobenzotrifluoride
PubChem CID 2737535
IUPAC Name 1-[4-fluoro-2-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(F)C=C1C(F)(F)F

Specifications

Refractive Index 1.449
Quantity 5 g
Beilstein 7637196
Formula Weight 206.14
Percent Purity 97%
Chemical Name or Material 4'-Fluoro-2'-(trifluoromethyl)acetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'