missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)phenol, 95%, Thermo Scientific Chemicals

Product Code. 15454337
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130047-19-7
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD00061290
InChI Key RNOVGJWJVRESAA-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl phenol, 2-trifluoromethyl-4-fluorophenol, 5-fluoro-2-hydroxybenzotrifluoride, pubchem4146, acmc-1bxww, intermediates-zcf02005, ksc495m8f, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl-phenol
PubChem CID 518675
IUPAC Name 4-fluoro-2-(trifluoromethyl)phenol
SMILES OC1=C(C=C(F)C=C1)C(F)(F)F

Specifications

Melting Point 50°C to 54°C
Quantity 1 g
UN Number UN3261
Formula Weight 180.1
Percent Purity 95%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'