missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3,5-dimethylbenzyl bromide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Fluoro-3,5-dimethylbenzyl bromide is used as laboratory chemical. It acts as an intermediate in the pharmaceutical industry.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
4-Fluoro-3,5-dimethylbenzyl bromide is used as laboratory chemical. It acts as an intermediate in the pharmaceutical industry.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | 44°C to 46°C |
Quantity | 1 g |
UN Number | UN3261 |
Formula Weight | 217.08 |
Percent Purity | 97% |
Chemical Name or Material | 4-Fluoro-3,5-dimethylbenzyl bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'