missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methyl(thiobenzamide), 97%, Thermo Scientific™

Product Code. 15416637
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1016775-44-2
Molecular Formula C8H8FNS
Molecular Weight (g/mol) 169.217
MDL Number MFCD09757567
InChI Key GTDSYOIDNUFACN-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylbenzene-1-carbothioamide, 4-fluoro-3-methyl thiobenzamide, 4-fluoro-3-methyl-thiobenzamide
PubChem CID 24695488
IUPAC Name 4-fluoro-3-methylbenzenecarbothioamide
SMILES CC1=C(C=CC(=C1)C(=S)N)F

Specifications

Melting Point 89°C to 91°C
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methyl(thiobenzamide)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'