missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™

Product Code. 10821681
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82297-89-0, 109-99-9
CAS Min % 8.53, 87.21
CAS Max % 12.79, 91.48
Color Brown to Yellow
Density 1.0010g/mL
Flash Point −26°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C7H6BrFMg
Linear Formula F(CH3)C6H3MgBr
MDL Number MFCD00075303
Quantity 50 mL
Specific Gravity 1.001
Synonym 4-fluoro-3-methylphenylmagnesium bromide, olsgdqispracfy-uhfffaoysa-m, bromo 4-fluoro-3-methylphenyl magnesium, 3-methyl-4-fluorophenyl magnesium bromide, 4-fluoro-3-methylphenyl magnesium bromide, 4-fluoro-3-methylphenylmagnesium bromide, 0.50 m in 2-methf, 4-fluoro-3-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 4-fluoro-3-methylphenylmagnesium bromide solution, 1.0 m in thf, grignard reagent
InChI Key OLSGDQISPRACFY-UHFFFAOYSA-M
SMILES CC1=CC([Mg]Br)=CC=C1F
IUPAC Name magnesium;1-fluoro-2-methylbenzene-4-ide;bromide
Molecular Weight (g/mol) 213.33
PubChem CID 2778169
Formula Weight 213.33
Physical Form Solution
Chemical Name or Material 4-Fluoro-3-methylphenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Fluoro-3-methylphenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.