missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitroanisole, 95%, Thermo Scientific Chemicals

Product Code. 15448118
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61324-93-4
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.127
MDL Number MFCD11848442
InChI Key ZRIKJXDEJYMBEJ-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitroanisole, benzene, 1-fluoro-4-methoxy-2-nitro, 3-fluoro-4-nitrobenzene,-1-methoxy, ksc613a5f, 2-fluoro-5-methoxynitrobenzene, 2-fluoro-5-methoxy-nitrobenzene, #
PubChem CID 565653
IUPAC Name 1-fluoro-4-methoxy-2-nitrobenzene
SMILES COC1=CC(=C(C=C1)F)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 171.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-nitroanisole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'