Learn More
4-Fluoro-3-nitrobenzonitrile, 97%, Thermo Scientific Chemicals
£55.50 - £196.00
Chemical Identifiers
CAS | 1009-35-4 |
---|---|
Molecular Formula | C7H3FN2O2 |
Molecular Weight (g/mol) | 166.111 |
MDL Number | MFCD01632197 |
InChI Key | LKOWKPGBAZVHOF-UHFFFAOYSA-N |
Synonym | 3-nitro-4-fluorobenzonitrile, 4-fluoro-3-nitro-benzonitrile, 5-cyano-2-fluoronitrobenzene, 4-cyano-1-fluoro-2-nitrobenzene, benzonitrile, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenecarbonitrile, 5-cyano-5-fluoro-1-nitrobenzene, pubchem4796, acmc-1c2vd, 4-fluro-3-nitrobenzonitrile |
PubChem CID | 2774654 |
IUPAC Name | 4-fluoro-3-nitrobenzonitrile |
SMILES | C1=CC(=C(C=C1C#N)[N+](=O)[O-])F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15415117
|
Thermo Scientific Alfa Aesar
H32391.03 |
1 g |
£55.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15425117
|
Thermo Scientific Alfa Aesar
H32391.06 |
5 g |
£196.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Chemical Identifiers
1009-35-4 | |
166.111 | |
LKOWKPGBAZVHOF-UHFFFAOYSA-N | |
2774654 | |
C1=CC(=C(C=C1C#N)[N+](=O)[O-])F |
C7H3FN2O2 | |
MFCD01632197 | |
3-nitro-4-fluorobenzonitrile, 4-fluoro-3-nitro-benzonitrile, 5-cyano-2-fluoronitrobenzene, 4-cyano-1-fluoro-2-nitrobenzene, benzonitrile, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenecarbonitrile, 5-cyano-5-fluoro-1-nitrobenzene, pubchem4796, acmc-1c2vd, 4-fluro-3-nitrobenzonitrile | |
4-fluoro-3-nitrobenzonitrile |
Specifications
1009-35-4 | |
C7H3FN2O2 | |
UN3439 | |
Insoluble in water. | |
C1=CC(=C(C=C1C#N)[N+](=O)[O-])F | |
166.111 | |
166.11 | |
4-Fluoro-3-nitrobenzonitrile |
90°C to 94°C | |
MFCD01632197 | |
3-nitro-4-fluorobenzonitrile, 4-fluoro-3-nitro-benzonitrile, 5-cyano-2-fluoronitrobenzene, 4-cyano-1-fluoro-2-nitrobenzene, benzonitrile, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenecarbonitrile, 5-cyano-5-fluoro-1-nitrobenzene, pubchem4796, acmc-1c2vd, 4-fluro-3-nitrobenzonitrile | |
LKOWKPGBAZVHOF-UHFFFAOYSA-N | |
4-fluoro-3-nitrobenzonitrile | |
2774654 | |
97% |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only