missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-phenoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 15447708
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68359-57-9
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.21
MDL Number MFCD01318148
InChI Key JDICMOLUAHZVDS-UHFFFAOYSA-N
Synonym 4-fluoro-3-phenoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-phenoxy, m-phenoxy-p-fluorobenzaldehyde, 2-fluoro-5-formyldiphenyl ether, 4-fluoro-3-phenoxy benzaldehyde, pubchem14130, 3-phenoxy-4fluorobenzaldehyde, ksc497q9b, 3-phenoxy-4-fluorobenzaldehyde, 3-phenoxy-4-fluoro-benzaldehyde
PubChem CID 110068
IUPAC Name 4-fluoro-3-phenoxybenzaldehyde
SMILES FC1=CC=C(C=O)C=C1OC1=CC=CC=C1

Specifications

Color Colorless
Density 1.209 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.583
Quantity 25 g
UN Number UN3082
Formula Weight 216.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-phenoxybenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'