missing translation for 'onlineSavingsMsg'
Learn More
4'-Fluoro-3'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™ Product Code.: 11487817

4'-Fluoro-3'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 11487817
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11487817

Brand: Thermo Scientific Alfa Aesar A16501.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208173-24-4
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD00061252
InChI Key SYFHRXQPXHETEF-UHFFFAOYSA-N
Synonym 4'-fluoro-3'-trifluoromethyl acetophenone, 1-4-fluoro-3-trifluoromethyl phenyl ethanone, 4-fluoro-3-trifluoromethyl acetophenone, 4'-fluoro-3'-trifluoromethylacetophenone, 1-4-fluoro-3-trifluoromethyl phenyl ethan-1-one, 5-acetyl-2-fluorobenzotrifluoride, ethanone, 1-4-fluoro-3-trifluoromethyl phenyl, 1-acetyl-4-fluoro-3-trifluoromethyl benzene, pubchem4321Show MoreShow Less
PubChem CID 605678
IUPAC Name 1-[4-fluoro-3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(F)C(=C1)C(F)(F)F

Specifications

Refractive Index 1.454
Quantity 1 g
Formula Weight 206.14
Percent Purity 97%
Chemical Name or Material 4'-Fluoro-3'-(trifluoromethyl)acetophenone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH18Q20 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10143047 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10097241 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10112355 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberH23Q23 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-Fluoro-3'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.