missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-5-nitrobenzene-1,2-diamine, Technical Grade, Thermo Scientific™

Product Code. 10200063
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 113269-06-0
Molecular Formula C6H6FN3O2
Molecular Weight (g/mol) 171.131
InChI Key QZNALIMEKAACKP-UHFFFAOYSA-N
PubChem CID 3782733
IUPAC Name 4-fluoro-5-nitrobenzene-1,2-diamine
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])F)N)N

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Synonym 1,2-benzenediamine,4-fluoro-5-nitro, 1,2-benzenediamine, 4-fluoro-5-nitro, acmc-20ag30, 4,5-diamine-2-fluoronitrobenzene, 5-fluoro-4-nitrobenzene-1,2-diamine, 4-fluoro-5-nitro-1,2-phenylenediamine
Formula Weight 171.13
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-Fluoro-5-nitrobenzene-1, 2-diamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'