missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-L-phenylalanine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.
Solubility
Soluble in water and 0.5M HCl (50 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.
Solubility
Soluble in water and 0.5M HCl (50 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | ∽240°C (decomposition) |
Beilstein | 2416148 |
Solubility Information | Soluble in water and 0.5M HCl (50mg/ml). |
Formula Weight | 183.18 |
Percent Purity | ≥98% |
Quantity | 1 g |
Optical Rotation | −25° (c=1 in Water) |
Chemical Name or Material | 4-Fluoro-L-phenylalanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'