missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-(4-fluorobenzyl)benzamide, 97%, Thermo Scientific™

Product Code. 15494148
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512181-73-6
Molecular Formula C14H11F2NO
Molecular Weight (g/mol) 247.245
InChI Key CJFYBDORFMDXTG-UHFFFAOYSA-N
Synonym 4-fluoro-n-4-fluorophenyl methyl benzamide, 4-fluoro-n-4-fluorobenzyl benzamide, n-4-fluorobenzyl-4-fluorobenzamide, 4-fluoro-n-4-fluoro-benzyl-benzamide
PubChem CID 2303704
IUPAC Name 4-fluoro-N-[(4-fluorophenyl)methyl]benzamide
SMILES C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)F)F

Specifications

Quantity 250 mg
Formula Weight 247.24
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-(4-fluorobenzyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'