missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzamide, 98%, Thermo Scientific Chemicals

Product Code. 11347006
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-75-9
Molecular Formula C7H6FNO
Molecular Weight (g/mol) 139.129
MDL Number MFCD00007992
InChI Key VNDHYTGVCGVETQ-UHFFFAOYSA-N
Synonym p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407
PubChem CID 71572
IUPAC Name 4-fluorobenzamide
SMILES C1=CC(=CC=C1C(=O)N)F

Specifications

Melting Point 154°C to 157°C
Quantity 100 g
Beilstein 1859744
Formula Weight 139.13
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'