missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoronaphthalene-1-carbonitrile, 97%

Product Code. 11431010
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11431010

Brand: Thermo Scientific Alfa Aesar B25335.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13916-99-9 | C11H6FN | 171.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13916-99-9
Molecular Formula C11H6FN
Molecular Weight (g/mol) 171.174
MDL Number MFCD00236656
InChI Key XQSGYYNFKIQWAC-UHFFFAOYSA-N
PubChem CID 2736734
IUPAC Name 4-fluoronaphthalene-1-carbonitrile
SMILES C1=CC=C2C(=C1)C(=CC=C2F)C#N

Specifications

Melting Point 89°C to 91°C
Quantity 5 g
UN Number UN3439
Synonym 1-cyano-4-fluoronaphthalene, 4-fluoro-1-naphthonitrile, 4-fluoro-1-naphthalenecarbonitrile, 1-naphthalenecarbonitrile,4-fluoro, 1-naphthalenecarbonitrile, 4-fluoro, 4-fluoronaphthalenecarbonitrile, 4-fluoro-1-phthonitrile, acmc-1buk6, 1-cyano-4-fluoronapthalene, 4-cyano-1-fluoronaphthalene
Formula Weight 171.18
Percent Purity 97%
Chemical Name or Material 4-Fluoronaphthalene-1-carbonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.