missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenethylealcohol, 97%, ACROS Organics™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Colorless |
Density | 1.1200g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | FC6H4CH2CH2OH |
Refractive Index | 1.5060 to 1.5080 (20°C, 589nm) |
Quantity | 25 g |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'