missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenol, 99%, Thermo Scientific Chemicals

Product Code. 11473944
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Fluorophenol, 99%, C6H5FO, CAS Number-371-41-5, ccris 665, fpn, 4-fluoro-phenol, p-fluorophenol, phenol, p-fluoro, para-fluorophenol, phenol, 4-fluoro, 4-fluoro phenol, 4-fluoranylphenol, 500g, 1362752, 185 deg.C to 188 deg.C, 1.22, Hazard Class: 6.1; Packaging Group: III, 206-736-0

4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.

Solubility
Soluble in water (80 mg/ml at 20°C), methylene dichloride, chloroform and methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-41-5
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002316
InChI Key RHMPLDJJXGPMEX-UHFFFAOYSA-N
Synonym p-fluorophenol, phenol, 4-fluoro, phenol, p-fluoro, para-fluorophenol, 4-fluoro phenol, 4-fluoro-phenol, ccris 665, 4-fluoranylphenol, fpn
PubChem CID 9732
IUPAC Name 4-fluorophenol
SMILES OC1=CC=C(F)C=C1

Specifications

Melting Point 44°C to 50°C
Density 1.22
Boiling Point 185°C to 188°C
Flash Point 68°C (154°F)
Odor Phenol-like
Quantity 500 g
UN Number UN2923
Beilstein 1362752
Solubility Information Soluble in water (80mg/ml at 20°C),methylene dichloride,chloroform and methanol.
Formula Weight 112.1
Percent Purity 99%
Chemical Name or Material 4-Fluorophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'