missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Fluorophenyl)methanesulfonyl chloride, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 208.64 |
Percent Purity | 95% |
Chemical Name or Material | (4-Fluorophenyl)methanesulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'