missing translation for 'onlineSavingsMsg'
Learn More

(4-Fluorophenyl)methanesulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10576694
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 103360-04-9
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD01631930
InChI Key UUQGWVIRPCRTSA-UHFFFAOYSA-N
Synonym 4-fluorophenyl methanesulfonyl chloride, 4-fluorobenzylsulfonyl chloride, 4-fluoro-phenyl-methanesulfonyl chloride, 4-fluorophenyl methyl sulfonyl chloride, chloro 4-fluorophenyl methyl sulfone, 4-fluorophenyl methanesulphonyl chloride, 4-fluoro-alpha-toluenesulfonyl chloride, benzenemethanesulfonyl chloride, 4-fluoro, 4-fluoro-phenyl-methane sulfonyl chloride, pubchem10056
PubChem CID 2759108
IUPAC Name (4-fluorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 208.64
Percent Purity 95%
Chemical Name or Material (4-Fluorophenyl)methanesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.