missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetone, 99%, Thermo Scientific Chemicals

£25.23 - £80.30

Chemical Identifiers

CAS 459-03-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00000362
InChI Key ZUEKIIWSVFBTCM-UHFFFAOYSA-N
Synonym 4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one
PubChem CID 521187
IUPAC Name 1-(4-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(F)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342587
SDS View Documents
Thermo Scientific Alfa Aesar
L11720.03
1 g
£25.23
1g
Please to purchase this item. Need a web account? Register with us today!
11352587
SDS View Documents
Thermo Scientific Alfa Aesar
L11720.06
5 g
£80.30
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 207-284-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluorophenylacetone, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.