missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylhydrazine hydrochloride, 97%

Product Code. 11407517
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11407517

Brand: Thermo Scientific Alfa Aesar A15624.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 823-85-8 | C6H8ClFN2 | 162.59 g/mol

4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylhydrazine hydrochloride is used as a chemical intermediate in the pharmaceutica1ls particularly of tryptamine drugs used in the treatment of migraine and cluster headaches. It is used to prepare agrochemical, and other chemical. It is used to detect sugars and aldehydes in analytical chemistry.

Solubility
Soluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-85-8
Molecular Formula C6H8ClFN2
Molecular Weight (g/mol) 162.59
MDL Number MFCD00012942
InChI Key FEKUXLUOKFSMRO-UHFFFAOYSA-N
Synonym 4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride
PubChem CID 69981
IUPAC Name (4-fluorophenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=C(F)C=C1

Specifications

Melting Point ∼265°C (decomposition)
Quantity 25 g
Beilstein 3627721
Solubility Information Soluble in water.
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 4-Fluorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.